Learn More
MSC North by Honeywell Hypalon Dry Box Glove, 7 in. Port, 15 mil, 32 in. Length, 1 ea.
Catalog No. 5000508283
Encompass
These dry box gloves are designed for use in controlled environments requiring high chemical resistance and durability. Constructed from Hypalon, they provide excellent flexibility and strength for demanding applications.
  • Manufactured from chemical-resistant Hypalon material
  • Designed for use with 7 in. diameter ports
  • Measures 15 mil in thickness
  • Total length of 32 in.
  • Available in size 9.75
Catalog No. 50-005-08283 Supplier MSC Supplier No. 48275861
These dry box gloves are designed for use in controlled environments requiring high chemical resistance and durability. Constructed from Hypalon, they provide excellent flexibility and strength for demanding applications.
  • Manufactured from chemical-resistant Hypalon material
  • Designed for use with 7 in. diameter ports
  • Measures 15 mil in thickness
  • Total length of 32 in.
  • Available in size 9.75