Learn More
Medchemexpress LLC HY-N2179 5mg Medchemexpress, Hypaphorine CAS:487-58-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN21795MG
Encompass_Preferred
Medchemexpress, HY-N2179 5mg Hypaphorine CAS:487-58-1 Hypaphorine is an indole alkaloid isolated from Pisolithus tinctorius, and with neurological and glucose-lowering effects in rodents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2874