Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES SpectraComp Flex Multipeak, Neg + pos (50% high/50% med), 4-species, 50 tests
SDP

Catalog No. NC3055280
Encompass_Preferred

SpectraComp Flex Multipeak controls are precision engineered single stain cell mimics that provide two levels of brightness without requiring titration of antibodies that may be off-scale using single intensity SpectraComp.

Specifications:
Catalog Number: SL-00028-A
Format: Neg + pos (50% high/50% med), 4-species
Package Size: 50 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 18 months
Country of Origin: United States
For Research Use Only.

Catalog No. NC3055280 Supplier SLINGSHOT BIOSCIENCES Supplier No. SL00028A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SpectraComp Flex Multipeak controls are precision engineered single stain cell mimics that provide two levels of brightness without requiring titration of antibodies that may be off-scale using single intensity SpectraComp.

Specifications:
Catalog Number: SL-00028-A
Format: Neg + pos (50% high/50% med), 4-species
Package Size: 50 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 18 months
Country of Origin: United States
For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.