Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLINGSHOT BIOSCIENCES SpectraComp Flex XT, Separate pos/neg vials, 5-species, 50 tests
SDP

Catalog No. NC2542531
Encompass_Preferred

SpectraComp Flex XT controls are precision engineered single stain cell mimics designed to deliver accurate compensation and spectral unmixing, with separate positive and negative particles and expanded five species antibody compatibility for high parameter flow cytometry workflows.

Specifications:
Catalog Number: SSB-22-A
Format: Separate pos/neg vials, 5-species
Package Size: 50 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Catalog No. NC2542531 Supplier SLINGSHOT BIOSCIENCES Supplier No. SSB22A
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SpectraComp Flex XT controls are precision engineered single stain cell mimics designed to deliver accurate compensation and spectral unmixing, with separate positive and negative particles and expanded five species antibody compatibility for high parameter flow cytometry workflows.

Specifications:
Catalog Number: SSB-22-A
Format: Separate pos/neg vials, 5-species
Package Size: 50 tests
Storage Conditions: 2 - 8°C
Shipping Conditions: No Ice
Shelf Life: 12 months
Country of Origin: United States
For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.