Learn More
Apexbio Technology LLC Hyper Assembly Cloning Kit, 100 rxns. Cas: MFCD: NA. used for fast, directional cloning of one or more fragments of DNA into any vector.
SDP
Catalog No. 501998290
Encompass_Preferred
Hyper Assembly Cloning Kit is designed for fast, directional cloning of one or more target fragment of DNA into any vector. This method employs the 3-5 exonuclease activity of Hyper Assembly Enzyme to get 15-20nt complementary arms at the ends of PCR-generated inserts and linearized vectors, then fuse the DNA fragments to get the recombinant vectors. These 15-20-bp overlaps can be engineered by designing primers for amplification of the desired sequences. Clone any insert into any location within any vector you choose Efficiently clone a broad range of fragment sizes Clone multiple DNA fragments simultaneously into any vector in a single reaction No restriction digestion, phosphatase treatment, or ligation required Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8290 Supplier Apexbio Technology LLC Supplier No. K1054100
Hyper Assembly Cloning Kit is designed for fast, directional cloning of one or more target fragment of DNA into any vector. This method employs the 3-5 exonuclease activity of Hyper Assembly Enzyme to get 15-20nt complementary arms at the ends of PCR-generated inserts and linearized vectors, then fuse the DNA fragments to get the recombinant vectors. These 15-20-bp overlaps can be engineered by designing primers for amplification of the desired sequences. Clone any insert into any location within any vector you choose Efficiently clone a broad range of fragment sizes Clone multiple DNA fragments simultaneously into any vector in a single reaction No restriction digestion, phosphatase treatment, or ligation required Other sizes are also available. Please inqury us for quote.