Learn More
ATHENA ES Hyper Broth™ 500g
Catalog No. NC0819109
Encompass_Preferred
Hyper Broth™ is an APF Certified™ proprietary media formulation developed by AthenaES™ for improved expression of recombinant proteins in E. coli. The medium is a complex, rich formulation which supplies amino acids, vitamins, and glucose at high levels. This medium yields the highest level of biomass of all AthenaES™ media. The medium is buffered at pH 7.3 ± 0.2 and has the highest buffering capacity of Athena’s Expression Media, which permits high glucose levels. For expression of recombinant proteins, induction should be done at cell densities three times that of LB Broth. Cell yields are typically 5 times that of LB Broth depending on the strain. Plasmid copy numbers are significantly higher than they are for LB Broth. This medium is suitable as the base for fermentation cultures and is excellent for feed-batch applications. Biomass yields will depend on the process and the strain employed.
Catalog No. NC0819109 Supplier ATHENA ES Supplier No. 0107
Hyper Broth™ is an APF Certified™ proprietary media formulation developed by AthenaES™ for improved expression of recombinant proteins in E. coli. The medium is a complex, rich formulation which supplies amino acids, vitamins, and glucose at high levels. This medium yields the highest level of biomass of all AthenaES™ media. The medium is buffered at pH 7.3 ± 0.2 and has the highest buffering capacity of Athena’s Expression Media, which permits high glucose levels. For expression of recombinant proteins, induction should be done at cell densities three times that of LB Broth. Cell yields are typically 5 times that of LB Broth depending on the strain. Plasmid copy numbers are significantly higher than they are for LB Broth. This medium is suitable as the base for fermentation cultures and is excellent for feed-batch applications. Biomass yields will depend on the process and the strain employed.