Learn More
Glass Expansion Aluminum Hyper Skimmer Cone for PerkinElmer NexION 300/350/1000/1100/2000 (W1033995)
SDP
Catalog No. 501907690
Encompass_Preferred
Aluminum Hyper Skimmer Cone (W1033995) ¿ Standard hyper skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive samples, including HF, we recommend Platinum/Nickel Hyper Skimmer Cones, PE3015-Pt/Ni.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7690 Supplier Glass Expansion Supplier No. PE3015AL
Aluminum Hyper Skimmer Cone (W1033995) ¿ Standard hyper skimmer cone for PerkinElmer NexION 1000/2000/300/350 ICP-MS. Suitable for most common applications (e.g., routine aqueous samples, <5% acid matrix, non-HF, non-organic). Not recommended for samples containing HF. Use 10x Magnifier Inspection Tool with Built-In LED Illumination (NC2042261) to inspect cone orifice condition. When not in use, keep cones stored in original packaging for protection.For corrosive samples, including HF, we recommend Platinum/Nickel Hyper Skimmer Cones, PE3015-Pt/Ni.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001