Learn More
LUMIGEN INCORPORATED Lumigen HyPerBlu, 5 mL
Catalog No. NC0301422
Encompass
Lumigen HyPerBlu Chemiluminescent Reagent is a novel ready-to-use substrate for the direct detection of hydrogen peroxide. Reaction of the substrate with hydrogen peroxide rapidly generates sustained high-intensity luminescence for maximum detection sensitivity in solution assays. Coupled with oxidases the Lumigen HyPerBlu substrate also allows one step indirect quantitation of oxidase substrates or the oxidase itself.
Catalog No. NC0301422 Supplier LUMIGEN INCORPORATED Supplier No. HPB00005
Lumigen HyPerBlu Chemiluminescent Reagent is a novel ready-to-use substrate for the direct detection of hydrogen peroxide. Reaction of the substrate with hydrogen peroxide rapidly generates sustained high-intensity luminescence for maximum detection sensitivity in solution assays. Coupled with oxidases the Lumigen HyPerBlu substrate also allows one step indirect quantitation of oxidase substrates or the oxidase itself.