Learn More
CPC Scientific H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-NH2 (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110344
Encompass
ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTA-NH2MOLECULAR FORMULA: C180H288N58O47MOLECULAR WEIGHT:4016.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: N. Horvichi et al., Science, 238, 1566 (1987); B.E. Kemp et al., Science, 238, 1568 (1987)
Catalog No. 50-211-0344 Supplier CPC Scientific Supplier No. PTHP006B
ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTA-NH2MOLECULAR FORMULA: C180H288N58O47MOLECULAR WEIGHT:4016.6STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: N. Horvichi et al., Science, 238, 1566 (1987); B.E. Kemp et al., Science, 238, 1568 (1987)