Learn More
CPC Scientific H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-Glu-Ile-Arg-Ala-Thr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110351
Encompass
A parathyroid hormone (PTH)-related peptide (1-40), human.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAEIRATSMOLECULAR FORMULA: C207H334N66O58MOLECULAR WEIGHT:4675.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [120298-73-9], SYNONYMS: pTH-Related Protein (1-40) (human, mouse, rat)RESEARCH AREA: Osteoporosis
Catalog No. 50-211-0351 Supplier CPC Scientific Supplier No. PTHP012A
A parathyroid hormone (PTH)-related peptide (1-40), human.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAEIRATSMOLECULAR FORMULA: C207H334N66O58MOLECULAR WEIGHT:4675.34STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [120298-73-9], SYNONYMS: pTH-Related Protein (1-40) (human, mouse, rat)RESEARCH AREA: Osteoporosis