Learn More
CPC Scientific H-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-NH2 (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110345
Encompass
Potent antagonist of PTH-related protein (1-34) in vitro and in vivo.ONE-LETTER SEQUENCE: LLHDKGKSIQDLRRRFFLHHLIAEIHTA-NH2MOLECULAR FORMULA: C153H247N49O37MOLECULAR WEIGHT:3364.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [115695-30-2], SYNONYMS: pTHrP (7-34) amide (human, mouse, rat), pTH-Related Protein (7-34) amide (human, mouse, rat)RESEARCH AREA: OsteoporosisREFERENCES: R.L. McKee et al., J. Bone and Mineral Res., 3, S73 (1988)
Catalog No. 50-211-0345 Supplier CPC Scientific Supplier No. PTHP007A
Potent antagonist of PTH-related protein (1-34) in vitro and in vivo.ONE-LETTER SEQUENCE: LLHDKGKSIQDLRRRFFLHHLIAEIHTA-NH2MOLECULAR FORMULA: C153H247N49O37MOLECULAR WEIGHT:3364.95STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [115695-30-2], SYNONYMS: pTHrP (7-34) amide (human, mouse, rat), pTH-Related Protein (7-34) amide (human, mouse, rat)RESEARCH AREA: OsteoporosisREFERENCES: R.L. McKee et al., J. Bone and Mineral Res., 3, S73 (1988)