Learn More
CPC Scientific H-Ala-Val-Ser-Glu-His-Gln-Leu-Leu-His-Asp-Lys-Gly-Lys-Ser-Ile-Gln-Asp-Leu-Arg-Arg-Arg-Phe-Phe-Leu-His-His-Leu-Ile-Ala-Glu-Ile-His-Thr-Ala-OH (trifluoroacetate salt) 0.5MG
SDP
Catalog No. 502110341
Encompass
Peptide found to have anabolic effect on bone formation in rats that can inhibit the rapid decline in bone formation due to denervation.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAMOLECULAR FORMULA: C180H287N57O48MOLECULAR WEIGHT:4016.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: L.J. Suva et al., Science, 237, 893 (1987)
Catalog No. 50-211-0341 Supplier CPC Scientific Supplier No. PTHP005A
Peptide found to have anabolic effect on bone formation in rats that can inhibit the rapid decline in bone formation due to denervation.ONE-LETTER SEQUENCE: AVSEHQLLHDKGKSIQDLRRRFFLHHLIAEIHTAMOLECULAR FORMULA: C180H287N57O48MOLECULAR WEIGHT:4016.57STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [112955-31-4], RESEARCH AREA: OsteoporosisREFERENCES: L.J. Suva et al., Science, 237, 893 (1987)