Learn More
Apexbio Technology LLC HyperChrom SP HP Agarose 100ml
SDP
Catalog No. 5000064363
Encompass_Preferred
HyperChrom SP HP Agarose is a strong cation-exchange chromatography resin designed for bioseparation applications based on differences in molecular charge properties Employing a highly cross-linked agarose matrix this resin enables efficient separation of biological molecules such as recombinant proteins antibodies nucleic acids viruses virus-like particles and polysaccharides The chemically modified agarose backbone enhances rigidity allowing high flow rates under reduced backpressure facilitating streamlined scalable purification processes suitable for biomedical research and biopharmaceutical production
Catalog No. 50-000-64363 Supplier Apexbio Technology LLC Supplier No. PC2015100
HyperChrom SP HP Agarose is a strong cation-exchange chromatography resin designed for bioseparation applications based on differences in molecular charge properties Employing a highly cross-linked agarose matrix this resin enables efficient separation of biological molecules such as recombinant proteins antibodies nucleic acids viruses virus-like particles and polysaccharides The chemically modified agarose backbone enhances rigidity allowing high flow rates under reduced backpressure facilitating streamlined scalable purification processes suitable for biomedical research and biopharmaceutical production