Learn More
Apexbio Technology LLC HyperFluor™ 488 Rabbit Anti-Goat IgG (H+L) Antibody 2X1ml
SDP
Catalog No. 5000097034
Encompass_Preferred
HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody is a fluorescently labeled secondary antibody targeting goat immunoglobulins (IgG heavy and light chains) It is designed to bind specifically to the Fc and light chain regions of goat antibodies thereby enabling detection of goat-derived primary antibodies in various immunoassays HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity through selective recognition of goat IgG by affinity-purified polyclonal rabbit antibodies conjugated with the HyperFluor 488 fluorophore Based on these properties HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence flow cytometry Western blot and immunohistochemistry applications
Catalog No. 50-000-97034 Supplier Apexbio Technology LLC Supplier No. K12142
HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody is a fluorescently labeled secondary antibody targeting goat immunoglobulins (IgG heavy and light chains) It is designed to bind specifically to the Fc and light chain regions of goat antibodies thereby enabling detection of goat-derived primary antibodies in various immunoassays HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity through selective recognition of goat IgG by affinity-purified polyclonal rabbit antibodies conjugated with the HyperFluor 488 fluorophore Based on these properties HyperFluor 488 Rabbit Anti-Goat IgG HplusL Antibody holds research potential in immunofluorescence flow cytometry Western blot and immunohistochemistry applications