Learn More
Apexbio Technology LLC HyperFluor™ 555 NHS Ester 1mg
SDP
Catalog No. 5000092144
Encompass_Preferred
HyperFluor 555 NHS Ester is a fluorescent labeling reagent targeting primary amines (R NH ) on biomolecules It is designed to facilitate covalent conjugation with proteins amine-modified oligonucleotides and other primary amine-containing biomolecules via its NHS ester moiety HyperFluor 555 NHS Ester exerts its activity through amine-specific covalent labeling Based on these properties HyperFluor 555 NHS Ester holds research potential in fluorescence microscopy and flow cytometry for labeling and detection of proteins and oligonucleotides
Catalog No. 50-000-92144 Supplier Apexbio Technology LLC Supplier No. A80991
HyperFluor 555 NHS Ester is a fluorescent labeling reagent targeting primary amines (R NH ) on biomolecules It is designed to facilitate covalent conjugation with proteins amine-modified oligonucleotides and other primary amine-containing biomolecules via its NHS ester moiety HyperFluor 555 NHS Ester exerts its activity through amine-specific covalent labeling Based on these properties HyperFluor 555 NHS Ester holds research potential in fluorescence microscopy and flow cytometry for labeling and detection of proteins and oligonucleotides