Learn More
Apexbio Technology LLC HyperFluor™ 555 Rabbit Anti-Goat IgG (H+L) Antibody 5X100ul
SDP
Catalog No. 5000097073
Encompass_Preferred
HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal antibody reagent targeting goat immunoglobulin G (IgG) recognizing both heavy and light chains It is designed to detect and bind goat primary antibodies enabling signal amplification and visualization HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody mediates its activity by specific binding to goat IgG and is conjugated with HyperFluor 555 fluorophore (excitation 555 nm emission 568 nm) facilitating fluorescence-based detection Based on these characteristics HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody is applicable in immunofluorescence immunocytochemistry western blotting and flow cytometry
Catalog No. 50-000-97073 Supplier Apexbio Technology LLC Supplier No. K33120.5
HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody is a polyclonal antibody reagent targeting goat immunoglobulin G (IgG) recognizing both heavy and light chains It is designed to detect and bind goat primary antibodies enabling signal amplification and visualization HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody mediates its activity by specific binding to goat IgG and is conjugated with HyperFluor 555 fluorophore (excitation 555 nm emission 568 nm) facilitating fluorescence-based detection Based on these characteristics HyperFluor 555 Rabbit Anti-Goat IgG HplusL Antibody is applicable in immunofluorescence immunocytochemistry western blotting and flow cytometry