Learn More
Apexbio Technology LLC HyperFluor™ 594 NHS Ester 295348-87-7 1mg
SDP
Catalog No. 5000088559
Encompass_Preferred
HyperFluor 594 NHS Ester (CAS 295348-87-7) is a fluorescent labeling reagent used for covalent conjugation to primary amine groups (R-NH2) on biomolecules such as proteins and amine-modified oligonucleotides It is designed to enable the formation of stable amide bonds via its succinimidyl ester (NHS ester) functionality when incubated with amine-containing targets under physiological to slightly alkaline conditions HyperFluor 594 NHS Ester displays its utility through efficient amine-specific bioconjugation Upon conjugation the resulting biomolecule-dye complexes exhibit excitation at approximately 590 nm and emission at approximately 617 nm Based on these properties HyperFluor 594 NHS Ester is suitable for applications in fluorescence microscopy and flow cytometry
Catalog No. 50-000-88559 Supplier Apexbio Technology LLC Supplier No. A81101
HyperFluor 594 NHS Ester (CAS 295348-87-7) is a fluorescent labeling reagent used for covalent conjugation to primary amine groups (R-NH2) on biomolecules such as proteins and amine-modified oligonucleotides It is designed to enable the formation of stable amide bonds via its succinimidyl ester (NHS ester) functionality when incubated with amine-containing targets under physiological to slightly alkaline conditions HyperFluor 594 NHS Ester displays its utility through efficient amine-specific bioconjugation Upon conjugation the resulting biomolecule-dye complexes exhibit excitation at approximately 590 nm and emission at approximately 617 nm Based on these properties HyperFluor 594 NHS Ester is suitable for applications in fluorescence microscopy and flow cytometry