Learn More
Apexbio Technology LLC HyperFluor™ 594 Rabbit Anti-Goat IgG (H+L) Antibody 100ul
SDP
Catalog No. 5000097150
Encompass_Preferred
HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is a fluorophore-conjugated secondary antibody targeting goat IgG heavy and light chains It is designed to specifically bind to goat-derived primary antibodies facilitating detection in immunoassays HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity by recognizing and binding to goat IgG enabling fluorescent labeling with emission at 617 nm upon excitation at 590 nm Based on these properties HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is applicable in immunocytochemistry immunohistochemistry and flow cytometry
Catalog No. 50-000-97150 Supplier Apexbio Technology LLC Supplier No. K33080.1
HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is a fluorophore-conjugated secondary antibody targeting goat IgG heavy and light chains It is designed to specifically bind to goat-derived primary antibodies facilitating detection in immunoassays HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody exerts its biological activity by recognizing and binding to goat IgG enabling fluorescent labeling with emission at 617 nm upon excitation at 590 nm Based on these properties HyperFluor 594 Rabbit Anti-Goat IgG HplusL Antibody is applicable in immunocytochemistry immunohistochemistry and flow cytometry