Learn More
Medchemexpress LLC HY-116330A 1mg Medchemexpress, Hyperforin (dicyclohexylammonium salt) CAS:238074-03-8 Purity:>98%
SDP
Catalog No. 501969415
Encompass_Preferred
Medchemexpress, HY-116330A 1mg Hyperforin (dicyclohexylammonium salt) CAS:238074-03-8 Hyperforin dicyclohexylammonium salt (Hyperforin DCHA) is a transient receptor canonical 6 (TRPC6) channels activator. Hyperforin dicyclohexylammonium salt modulates Ca2+ levels by activating Ca2+-conducting non-selective canonical TRPC6 channels. Hyperforin dicyclohexylammonium salt shows antidepressant effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9415 Supplier Medchemexpress LLC Supplier No. HY116330A1MG
Medchemexpress, HY-116330A 1mg Hyperforin (dicyclohexylammonium salt) CAS:238074-03-8 Hyperforin dicyclohexylammonium salt (Hyperforin DCHA) is a transient receptor canonical 6 (TRPC6) channels activator. Hyperforin dicyclohexylammonium salt modulates Ca2+ levels by activating Ca2+-conducting non-selective canonical TRPC6 channels. Hyperforin dicyclohexylammonium salt shows antidepressant effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.