Learn More
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 500L Liner - Diameter: 31in. (79cm)
GREENER_CHOICE
Catalog No. SH3064707 Shop All Thermo Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
SH3064707 Non-sterile
1 options
Catalog No. SH3064707 Supplier Thermo Scientific™ Supplier No. SH30647.07
Add to Cart
Add to Cart
Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.
Standard Thermo Scientific tank liners feature an open top for maximum recovery, with drum dolly available to provide mobility.
Features:
  • Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
  • Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.
Applications of the Thermo Scientific tank liner:
  • Removes the need for tank cleaning and helps reduce cycle times.
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Outer Packaging Placed in strong cardboard cartons with product and lot identification.
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
For Use With (Equipment) Industry Standard Cylindrical Tanks
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9
Sterility Non-sterile