Learn More
Selleck Chemical LLC Hypericin S3273-5mg
SDP
Catalog No. 503110881
Encompass_Preferred
Hypericin (Hyp HY) is a naturally occurring substance found in the common St John s Wort (Hypericum species) with antidepressive antineoplastic and antiviral (human immunodeficiency and hepatitis C virus) activities Hypericin has inhibitory effects on MAO (monoaminoxidase) PKC (protein kinase C) dopamine-beta-hydroxylase reverse transcriptase telomerase and CYP (cytochrome P450)
Catalog No. 50-311-0881 Supplier Selleck Chemical LLC Supplier No. S32735MG
Hypericin (Hyp HY) is a naturally occurring substance found in the common St John s Wort (Hypericum species) with antidepressive antineoplastic and antiviral (human immunodeficiency and hepatitis C virus) activities Hypericin has inhibitory effects on MAO (monoaminoxidase) PKC (protein kinase C) dopamine-beta-hydroxylase reverse transcriptase telomerase and CYP (cytochrome P450)