Learn More
Apexbio Technology LLC Hypericin 548-04-9 10mg
SDP
Catalog No. 5000094460
Encompass_Preferred
Hypericin (CAS 548-04-9) is a small-molecule inhibitor targeting protein kinase C (PKC) and monoamine oxidase (MAO) It acts to selectively inhibit these enzymes thereby regulating cellular signal transduction pathways related to cell proliferation and apoptosis Hypericin exerts its biological activity primarily through selective inhibition of PKC In in vitro assays hypericin demonstrates an inhibitory profile with IC50 values against PKC ranging from approximately 100 to 250 nM depending on assay conditions Based on these pharmacological properties hypericin holds research potential in oncology virology and neuroscience
Catalog No. 50-000-94460 Supplier Apexbio Technology LLC Supplier No. N273910
Hypericin (CAS 548-04-9) is a small-molecule inhibitor targeting protein kinase C (PKC) and monoamine oxidase (MAO) It acts to selectively inhibit these enzymes thereby regulating cellular signal transduction pathways related to cell proliferation and apoptosis Hypericin exerts its biological activity primarily through selective inhibition of PKC In in vitro assays hypericin demonstrates an inhibitory profile with IC50 values against PKC ranging from approximately 100 to 250 nM depending on assay conditions Based on these pharmacological properties hypericin holds research potential in oncology virology and neuroscience