Learn More
Medchemexpress LLC Hypericin | 548-04-9 | 504.443 | 1 MG
SDP
Catalog No. 5000390137
Encompass_Preferred
Hypericin is a naturally occurring substance found in Hypericum perforatum L. It functions as an inhibitor of PKC (protein kinase C), MAO (monoaminoxidase), dopamine-beta-hydroxylase, reverse transcriptase, telomerase, and CYP (cytochrome P450). It exhibits antitumor, antiviral, and antidepressive activities, and can induce apoptosis.
  • Inhibits PKC (protein kinase C)
  • Inhibits MAO (monoaminoxidase)
  • Inhibits dopamine-beta-hydroxylase
  • Inhibits reverse transcriptase
  • Inhibits telomerase
  • Inhibits CYP (cytochrome P450)
  • Shows antitumor activity
  • Shows antiviral activity
  • Shows antidepressive activity
  • Induces apoptosis
  • Relevant for infection research
  • Relevant for neurological disease research
  • Relevant for cancer research
Catalog No. 50-003-90137 Supplier Medchemexpress LLC Supplier No. HYN04531MG
Hypericin is a naturally occurring substance found in Hypericum perforatum L. It functions as an inhibitor of PKC (protein kinase C), MAO (monoaminoxidase), dopamine-beta-hydroxylase, reverse transcriptase, telomerase, and CYP (cytochrome P450). It exhibits antitumor, antiviral, and antidepressive activities, and can induce apoptosis.
  • Inhibits PKC (protein kinase C)
  • Inhibits MAO (monoaminoxidase)
  • Inhibits dopamine-beta-hydroxylase
  • Inhibits reverse transcriptase
  • Inhibits telomerase
  • Inhibits CYP (cytochrome P450)
  • Shows antitumor activity
  • Shows antiviral activity
  • Shows antidepressive activity
  • Induces apoptosis
  • Relevant for infection research
  • Relevant for neurological disease research
  • Relevant for cancer research