Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hypericin | 548-04-9 | 98.0% | 504.44 g/mol | C30H16O8 | 10 MG
SDP

Catalog No. 5000295656
Encompass_Preferred

Hypericin is a photosensitive natural compound (C30H16O8) used in biochemical research as an inhibitor of protein kinase C (PKC) and monoamine oxidase (MAO). It is applied in studies of apoptosis, phototoxicity, antiviral activity, and antidepressant mechanisms, and is supplied as a solid or solution for in vitro experiments.

  • Used as a PKC and MAO inhibitor in biochemical research.
  • CAS number 548-04-9; molecular weight 504.44 g/mol.
  • Purity typically 98.0% and appears as a brown to black solid.
  • Soluble in DMSO (≈250 mg/mL with warming and ultrasonication).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); solutions -80°C (up to 6 months).
  • Available in small research quantities such as 10 MG and solution formats.

Catalog No. 50-002-95656 Supplier Medchemexpress LLC Supplier No. HYN045310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hypericin is a photosensitive natural compound (C30H16O8) used in biochemical research as an inhibitor of protein kinase C (PKC) and monoamine oxidase (MAO). It is applied in studies of apoptosis, phototoxicity, antiviral activity, and antidepressant mechanisms, and is supplied as a solid or solution for in vitro experiments.

  • Used as a PKC and MAO inhibitor in biochemical research.
  • CAS number 548-04-9; molecular weight 504.44 g/mol.
  • Purity typically 98.0% and appears as a brown to black solid.
  • Soluble in DMSO (≈250 mg/mL with warming and ultrasonication).
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); solutions -80°C (up to 6 months).
  • Available in small research quantities such as 10 MG and solution formats.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.