Learn More
Medchemexpress LLC Hypericin | 548-04-9 | MFCD00016683 | 98.2% | 504.44 | C30H16O8 | 10 MG
SDP
Catalog No. 5000296266
Encompass_Preferred
Hypericin is an analytical reference standard of a naturally occurring compound found in Hypericum perforatum. It is a brown to reddish-brown solid (C30H16O8; MW 504.44 g/mol) supplied at high purity for research and analytical applications, including qualitative and quantitative methods. Store and handle according to the Certificate of Analysis and applicable safety data.
  • Analytical reference standard suitable for HPLC, GC, and MS
  • High purity for reliable qualitative and quantitative analysis
  • Brown to reddish brown solid form for ease of handling in small-batch assays
  • Derived from plant source (Hypericum perforatum)
  • Supplied in small sizes appropriate for analytical work
Catalog No. 50-002-96266 Supplier Medchemexpress LLC Supplier No. HYN0453R10MG
Hypericin is an analytical reference standard of a naturally occurring compound found in Hypericum perforatum. It is a brown to reddish-brown solid (C30H16O8; MW 504.44 g/mol) supplied at high purity for research and analytical applications, including qualitative and quantitative methods. Store and handle according to the Certificate of Analysis and applicable safety data.
  • Analytical reference standard suitable for HPLC, GC, and MS
  • High purity for reliable qualitative and quantitative analysis
  • Brown to reddish brown solid form for ease of handling in small-batch assays
  • Derived from plant source (Hypericum perforatum)
  • Supplied in small sizes appropriate for analytical work