Learn More
TARGETMOL CHEMICALS INC HYPERICIN 5MG
SDP
Catalog No. 502576382
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Hypericin (Cyclosan) has antineoplastic activity. 2. Hypericin has antibiotic and antiviral activities. 3. Hypericin has antidepressive activity through inhibiting monoamine oxidase enzyme. 4. Hypericin inhibits RANKL-mediated osteoclastogenesis via affecting ERK signalling in vitro and suppresses wear particle-induced osteolysis in vivo. purity: 99%
Catalog No. 50-257-6382 Supplier TARGETMOL CHEMICALS INC Supplier No. T6S09235MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. 1. Hypericin (Cyclosan) has antineoplastic activity. 2. Hypericin has antibiotic and antiviral activities. 3. Hypericin has antidepressive activity through inhibiting monoamine oxidase enzyme. 4. Hypericin inhibits RANKL-mediated osteoclastogenesis via affecting ERK signalling in vitro and suppresses wear particle-induced osteolysis in vivo. purity: 99%