Learn More
Medchemexpress LLC Hypericin-d10 | 514.50 | 1 MG
SDP
Catalog No. 5000389775
Encompass_Preferred
Hypericin-d10 is the deuterium-labeled Hypericin. Hypericin is a photosensitive antiviral with anticancer and antidepressant properties, derived from *Hypericum perforatum*. It can inhibit tyrosine kinases with an IC50 of 7.5 μM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-89775 Supplier Medchemexpress LLC Supplier No. HYN0453S1MG
Hypericin-d10 is the deuterium-labeled Hypericin. Hypericin is a photosensitive antiviral with anticancer and antidepressant properties, derived from *Hypericum perforatum*. It can inhibit tyrosine kinases with an IC50 of 7.5 μM.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.