Learn More
Enzo Life Sciences Hypericin (native) (10mg). CAS: 548-04-9
SDP
Supplier:  Enzo Life Sciences ALX350030M010
Encompass_Preferred
A potent and selective inhibitor of protein kinase C (IC50=3.4µM). Anti-viral and anti-retroviral actitity. Displays antineoplastic and antitumor actitities. Most activities require light activation. Induces apoptosis in neuroblastoma cells at 1µM or above (IC50=0.1µM at 29 mW/cm2 (tungsten light)). Inhibitition of other kinases (e.g. EGFR-PTK, CKII and MAP kinase). Bright red fluorescence emissino and photostability. Antidepressant. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), 100% ethanol, methanol, acetone, ethylmethylketone, pyridine or other organic solvents and bases. Long Term Storage: +4°C.
Catalog No. 50-200-8699