Learn More
Meridian Life Science HYPERLADDER I-200 LANES
SDP
Catalog No. NC9841467
Encompass
HyperLadder I,quantitative,200 Lanes,Features: -- Each band represents an exact amount of dsDNA -- Supplied in ready-to-use form,5microliter per lane (720ng of DNA) -- Supplied with a 5x sample loading buffer,Applications -- Provides
Catalog No. NC9841467 Supplier Meridian Life Science Supplier No. BIO33025
HyperLadder I,quantitative,200 Lanes,Features: -- Each band represents an exact amount of dsDNA -- Supplied in ready-to-use form,5microliter per lane (720ng of DNA) -- Supplied with a 5x sample loading buffer,Applications -- Provides