Learn More
Meridian Life Science HYPERLADDER-II 200 LANES
SDP
Catalog No. NC9061542
Encompass
HyperLadder II, Quantitative, Size: 200 Lanes, Contains 15 regularly-spaced bands ranging from 50bp to 2kb, Each band contains an exact amount of DNA, enabling accurate quantitation. 5uL per lanegives a total of 600ng DNA
Catalog No. NC9061542 Supplier Meridian Life Science Supplier No. BIO33039
HyperLadder II, Quantitative, Size: 200 Lanes, Contains 15 regularly-spaced bands ranging from 50bp to 2kb, Each band contains an exact amount of DNA, enabling accurate quantitation. 5uL per lanegives a total of 600ng DNA