Learn More
Meridian Life Science HYPERLADDER IV-200 LANES
SDP
Catalog No. NC9977703
Encompass
Hyperladder IV, DNA Ladder 25bp to 1,000bp, 200 lanes
Catalog No. NC9977703 Supplier Meridian Life Science Supplier No. BIO33029
Hyperladder IV, DNA Ladder 25bp to 1,000bp, 200 lanes