Learn More
Meridian Life Science HYPERLADDER IV-500 LANES
SDP
Catalog No. NC9841468
Encompass
Hyperladder IV, quantitative, Size: 500 Lanes, Contains 9 regularly-spaced bands ranging from 100bp to 1kb, Each band contains an exact amount of DNA, Enabling accurate quantitation, 5microliter per lane gives a total of 500ng dsDNA
Catalog No. NC9841468 Supplier Meridian Life Science Supplier No. BIO33030
Hyperladder IV, quantitative, Size: 500 Lanes, Contains 9 regularly-spaced bands ranging from 100bp to 1kb, Each band contains an exact amount of DNA, Enabling accurate quantitation, 5microliter per lane gives a total of 500ng dsDNA