Learn More
Meridian Life Science HYPERLADDER V 100 LANE
SDP
Catalog No. NC9929754
Encompass
HyperLadder 25bp is a ready-to-use molecular weight marker for size determination of DNA fragments. It is especially designed for short fragments such as apoptotic DNA oligonucleotides. This ready-to-use format reduces handling steps.
Catalog No. NC9929754 Supplier Meridian Life Science Supplier No. BIO33057
HyperLadder 25bp is a ready-to-use molecular weight marker for size determination of DNA fragments. It is especially designed for short fragments such as apoptotic DNA oligonucleotides. This ready-to-use format reduces handling steps.