Learn More
Meridian Life Science HYPERLADDER V-200 LANES
SDP
Catalog No. NC9841469
Encompass
Hyperladder,200 lanes,12 bands, ranging from 25 to 500 bp,each band is an exact multiple of 25 bp and contains an exact amount of dsDNA, for DNA Size Determination and Quantification
Catalog No. NC9841469 Supplier Meridian Life Science Supplier No. BIO33031
Hyperladder,200 lanes,12 bands, ranging from 25 to 500 bp,each band is an exact multiple of 25 bp and contains an exact amount of dsDNA, for DNA Size Determination and Quantification