Learn More
TARGETMOL CHEMICALS INC HYPEROSIDE 25MG
SDP
Catalog No. 502580515
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hyperoside (Hyperin) is a naturally occuring flavonoid compound which shows multiple bioactivities including myocardial protection anti-redox and anti-inflammatory activities. purity: 99%
Catalog No. 50-258-0515 Supplier TARGETMOL CHEMICALS INC Supplier No. T284425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Hyperoside (Hyperin) is a naturally occuring flavonoid compound which shows multiple bioactivities including myocardial protection anti-redox and anti-inflammatory activities. purity: 99%