Learn More
Indofine Chemical Hyperoside With Hplc, 10 Mg
SDP
Supplier:  Indofine Chemical 021027S 10 MG
Encompass_Preferred
Molecular Formula: C21H20O12, Purity: 98% by HPLC, Molecular Weight: 464.38, Appearance: Yellow powder, Solubility: Soluble in Ethyl acetate:formic acid:acetic acid:water (100:11:11:27), Storage conditions: Dessicate, Also known as Hyperin, Quercetin-3-galactoside
Catalog No. 50-908-884