Learn More
Apexbio Technology LLC HyperScribe-T7-siRNA-Synth-Kit
SDP
Catalog No. 5000567572
Encompass_Preferred
HyperScribe T7 siRNA Synthesis Kit is a cutting-edge tool in the field of genetic and biomedical research designed for the efficient synthesis of small interfering RNA (siRNA) Originating from advanced molecular biology technologies this kit leverages the T7 RNA polymerase system to produce siRNA molecules that can specifically target and degrade complementary mRNA sequences thereby silencing gene expression The mechanism of action involves the RNA-induced silencing complex (RISC) pathway allowing precise gene regulation It exhibits potent in vitro activity often effective in nanomolar to low micromolar ranges Researchers commonly apply this kit in various experimental frameworks including cell models animal studies and drug screening initiatives to investigate gene function and therapeutic potentials Experimental concentrations typically depend on the specific research design ensuring flexibility and customization for diverse scientific inquiries
Catalog No. 50-005-67572 Supplier Apexbio Technology LLC Supplier No. K142250
HyperScribe T7 siRNA Synthesis Kit is a cutting-edge tool in the field of genetic and biomedical research designed for the efficient synthesis of small interfering RNA (siRNA) Originating from advanced molecular biology technologies this kit leverages the T7 RNA polymerase system to produce siRNA molecules that can specifically target and degrade complementary mRNA sequences thereby silencing gene expression The mechanism of action involves the RNA-induced silencing complex (RISC) pathway allowing precise gene regulation It exhibits potent in vitro activity often effective in nanomolar to low micromolar ranges Researchers commonly apply this kit in various experimental frameworks including cell models animal studies and drug screening initiatives to investigate gene function and therapeutic potentials Experimental concentrations typically depend on the specific research design ensuring flexibility and customization for diverse scientific inquiries