Learn More
Apexbio Technology LLC HyperScript™ III First-Strand cDNA Synthesis Kit 50rxn
SDP
Catalog No. 5000097198
Encompass_Preferred
HyperScript III First-Strand cDNA Synthesis Kit is an enzymatic reagent kit designed for generating first-strand complementary DNA (cDNA) from total RNA or poly(A) RNA templates It utilizes HyperScript III Reverse Transcriptase an engineered derivative of M-MLV reverse transcriptase with reduced RNase H activity and enhanced thermostability and fidelity HyperScript III First-Strand cDNA Synthesis Kit enables efficient synthesis of cDNA with extended length superior efficiency on GC-rich templates and compatibility with various primer types including random primers Oligo(dT) V/N and gene-specific primers Based on these features HyperScript III First-Strand cDNA Synthesis Kit is applicable for downstream applications such as PCR qPCR and gene expression analyses
Catalog No. 50-000-97198 Supplier Apexbio Technology LLC Supplier No. K158150
HyperScript III First-Strand cDNA Synthesis Kit is an enzymatic reagent kit designed for generating first-strand complementary DNA (cDNA) from total RNA or poly(A) RNA templates It utilizes HyperScript III Reverse Transcriptase an engineered derivative of M-MLV reverse transcriptase with reduced RNase H activity and enhanced thermostability and fidelity HyperScript III First-Strand cDNA Synthesis Kit enables efficient synthesis of cDNA with extended length superior efficiency on GC-rich templates and compatibility with various primer types including random primers Oligo(dT) V/N and gene-specific primers Based on these features HyperScript III First-Strand cDNA Synthesis Kit is applicable for downstream applications such as PCR qPCR and gene expression analyses