Learn More
Apexbio Technology LLC HyperScript™ III Reverse Transcriptase 2000U
SDP
Supplier:  Apexbio Technology LLC K31012000
Encompass_Preferred
HyperScript III Reverse Transcriptase is an engineered reverse transcriptase derived from Moloney Murine Leukemia Virus (M-MLV) designed to synthesize full-length cDNA from RNA targets Through reduced RNase H activity and enhanced thermal stability it efficiently transcribes RNA templates including those with complex secondary structures or high GC content into cDNA up to 12 3 kb in length with improved yield and accuracy This enzyme demonstrates robust cDNA synthesis activity and is supplied as a reaction-ready solution with a specialized buffer HyperScript III Reverse Transcriptase is suitable for applications such as quantitative PCR (qPCR) and transcriptome analysis
Catalog No. 50-000-97219