Learn More
Apexbio Technology LLC HyperScript™ III RT SuperMix for qPCR (with gDNA wiper) 50rxn
SDP
Catalog No. 5000097351
Encompass_Preferred
HyperScript III RT SuperMix for qPCR with gDNA wiper is an optimized reagent system designed for reverse transcription and genomic DNA removal in RT-qPCR applications It utilizes HyperScript III Reverse Transcriptase an engineered M-MLV derivative with reduced RNase H activity enhanced thermostability and improved fidelity to generate high-yield long cDNA strands efficiently including from GC-rich RNA templates The included gDNA wiper solution eliminates genomic DNA contamination prior to reverse transcription The kit contains a primer mix of Oligo(dT)23VN and random primers to ensure equal representation of different RNA regions HyperScript III RT SuperMix for qPCR with gDNA wiper is suitable for applications such as gene expression analysis using SYBR Green or probe-based qPCR adaptable for studies with low-copy gene targets
Catalog No. 50-000-97351 Supplier Apexbio Technology LLC Supplier No. K158550
HyperScript III RT SuperMix for qPCR with gDNA wiper is an optimized reagent system designed for reverse transcription and genomic DNA removal in RT-qPCR applications It utilizes HyperScript III Reverse Transcriptase an engineered M-MLV derivative with reduced RNase H activity enhanced thermostability and improved fidelity to generate high-yield long cDNA strands efficiently including from GC-rich RNA templates The included gDNA wiper solution eliminates genomic DNA contamination prior to reverse transcription The kit contains a primer mix of Oligo(dT)23VN and random primers to ensure equal representation of different RNA regions HyperScript III RT SuperMix for qPCR with gDNA wiper is suitable for applications such as gene expression analysis using SYBR Green or probe-based qPCR adaptable for studies with low-copy gene targets