Learn More
Apexbio Technology LLC HyperScript™ IV First-Strand cDNA Synthesis Kit 20rxn
SDP
Catalog No. 5000097200
Encompass_Preferred
HyperScript IV First-Strand cDNA Synthesis Kit contains an engineered M-MLV reverse transcriptase used for the synthesis of first-strand cDNA from RNA templates for molecular biology research The kit comprises HyperScript IV Reverse Transcriptase which is optimized for enhanced inhibitor tolerance faster elongation increased thermostability and reduced RNase H activity as well as two types of primers (Random Primer and Oligo(dT)23VN) to allow flexibility in experimental design Synthesized first-strand cDNA is suitable for downstream applications including PCR qPCR and other analytical approaches
Catalog No. 50-000-97200 Supplier Apexbio Technology LLC Supplier No. K158620
HyperScript IV First-Strand cDNA Synthesis Kit contains an engineered M-MLV reverse transcriptase used for the synthesis of first-strand cDNA from RNA templates for molecular biology research The kit comprises HyperScript IV Reverse Transcriptase which is optimized for enhanced inhibitor tolerance faster elongation increased thermostability and reduced RNase H activity as well as two types of primers (Random Primer and Oligo(dT)23VN) to allow flexibility in experimental design Synthesized first-strand cDNA is suitable for downstream applications including PCR qPCR and other analytical approaches