Learn More
Apexbio Technology LLC HyperScript™ IV First-Strand cDNA Synthesis SuperMix 50rxn
SDP
Supplier:  Apexbio Technology LLC K158750
Encompass_Preferred
HyperScript IV First-Strand cDNA Synthesis SuperMix is a reagent kit incorporating an engineered reverse transcriptase derived from M-MLV Reverse Transcriptase It is designed to efficiently catalyze first-strand cDNA synthesis from RNA templates thereby enabling reverse transcription required for downstream molecular analyses HyperScript IV First-Strand cDNA Synthesis SuperMix exhibits high activity enhanced inhibitor resistance extended elongation capability improved reaction kinetics and reduced RNase H activity supporting efficient full-length cDNA synthesis Synthesized cDNA is compatible with downstream applications such as PCR amplification and quantitative PCR (qPCR)
Catalog No. 50-000-97052