Learn More
Apexbio Technology LLC HyperScript™ IV First-Strand cDNA Synthesis SuperMix (with gDNA wiper) 50rxn
SDP
Catalog No. 5000097298
Encompass_Preferred
HyperScript IV First-Strand cDNA Synthesis SuperMix with gDNA wiper is a reverse transcription reagent system designed to synthesize first-strand cDNA from RNA templates The kit utilizes HyperScript IV Reverse Transcriptase a genetically engineered fourth-generation M-MLV-derived enzyme with enhanced inhibitor tolerance processivity and thermostability and reduced RNase H activity It incorporates a 4 gDNA wiper buffer for efficient removal of residual genomic DNA and offers flexible primer options including Random Primer and Oligo(dT)23VN Synthesized first-strand cDNA is suitable for downstream PCR or qPCR analyses Based on these properties HyperScript IV First-Strand cDNA Synthesis SuperMix with gDNA wiper holds research potential in gene expression studies and molecular biology applications
Catalog No. 50-000-97298 Supplier Apexbio Technology LLC Supplier No. K158950
HyperScript IV First-Strand cDNA Synthesis SuperMix with gDNA wiper is a reverse transcription reagent system designed to synthesize first-strand cDNA from RNA templates The kit utilizes HyperScript IV Reverse Transcriptase a genetically engineered fourth-generation M-MLV-derived enzyme with enhanced inhibitor tolerance processivity and thermostability and reduced RNase H activity It incorporates a 4 gDNA wiper buffer for efficient removal of residual genomic DNA and offers flexible primer options including Random Primer and Oligo(dT)23VN Synthesized first-strand cDNA is suitable for downstream PCR or qPCR analyses Based on these properties HyperScript IV First-Strand cDNA Synthesis SuperMix with gDNA wiper holds research potential in gene expression studies and molecular biology applications