Learn More
Apexbio Technology LLC HyperScript™ IV RT SuperMix for qPCR 100rxn
SDP
Catalog No. 5000097188
Encompass_Preferred
HyperScript IV RT SuperMix for qPCR is an engineered reverse transcriptase enzyme formula designed for efficient cDNA synthesis from RNA templates Developed from fourth-generation M-MLV Reverse Transcriptase it exhibits improved resistance to inhibitors enhanced processivity and optimized reaction kinetics while maintaining low RNase H activity and thermostability HyperScript IV RT SuperMix exerts its activity by catalyzing reverse transcription of RNA to cDNA using a balanced mix of Oligo(dT) 23VN primers random primers buffers dNTPs and cofactors It is suitable for two-step RT-qPCR procedures including those using low-concentration RNA samples Based on these properties HyperScript IV RT SuperMix for qPCR is suitable for research applications involving cDNA synthesis and quantitative PCR analysis
Catalog No. 50-000-97188 Supplier Apexbio Technology LLC Supplier No. K1588100
HyperScript IV RT SuperMix for qPCR is an engineered reverse transcriptase enzyme formula designed for efficient cDNA synthesis from RNA templates Developed from fourth-generation M-MLV Reverse Transcriptase it exhibits improved resistance to inhibitors enhanced processivity and optimized reaction kinetics while maintaining low RNase H activity and thermostability HyperScript IV RT SuperMix exerts its activity by catalyzing reverse transcription of RNA to cDNA using a balanced mix of Oligo(dT) 23VN primers random primers buffers dNTPs and cofactors It is suitable for two-step RT-qPCR procedures including those using low-concentration RNA samples Based on these properties HyperScript IV RT SuperMix for qPCR is suitable for research applications involving cDNA synthesis and quantitative PCR analysis