Learn More
Apexbio Technology LLC HyperScript™ IV RT SuperMix for qPCR (with gDNA wiper) 50rxn
SDP
Catalog No. 5000097352
Encompass_Preferred
HyperScript IV RT SuperMix for qPCR with gDNA wiper is a reagent system designed for efficient cDNA synthesis in two-step RT-qPCR applications It contains HyperScript IV Reverse Transcriptase an engineered variant of M-MLV Reverse Transcriptase with enhanced inhibitor tolerance elongation efficiency reaction kinetics and thermostability while maintaining low RNase H activity The kit employs a 4 gDNA wiper reagent to deplete genomic DNA prior to reverse transcription enabling contamination-free cDNA synthesis Resulting cDNA is compatible with both SYBR Green and probe-based qPCR assays This product is intended for research applications in accurate and consistent RNA quantification by RT-qPCR
Catalog No. 50-000-97352 Supplier Apexbio Technology LLC Supplier No. K159050
HyperScript IV RT SuperMix for qPCR with gDNA wiper is a reagent system designed for efficient cDNA synthesis in two-step RT-qPCR applications It contains HyperScript IV Reverse Transcriptase an engineered variant of M-MLV Reverse Transcriptase with enhanced inhibitor tolerance elongation efficiency reaction kinetics and thermostability while maintaining low RNase H activity The kit employs a 4 gDNA wiper reagent to deplete genomic DNA prior to reverse transcription enabling contamination-free cDNA synthesis Resulting cDNA is compatible with both SYBR Green and probe-based qPCR assays This product is intended for research applications in accurate and consistent RNA quantification by RT-qPCR