Learn More
Apexbio Technology LLC HYPERSCRIPT IV RTASE-2000mg
SDP
Catalog No. 5000897416
Encompass_Preferred
HyperScript IV Reverse Transcriptase is a pivotal enzyme in molecular biology research primarily used in the synthesis of complementary DNA (cDNA) from RNA templates Originating from retroviral sources this enzyme is crucial for studies involving gene expression analysis cloning and RNA sequencing It operates through a mechanism that reverses the transcription process effectively converting single-stranded RNA into stable cDNA Its activity is measured in the nanomolar to low micromolar range demonstrating high efficiency and reliability under in vitro conditions This enzyme is extensively applied in various experimental settings including cell models animal studies and high-throughput drug screening assays Experimental concentrations typically depend on the specific research design Using this advanced reverse transcriptase researchers can achieve precise and reproducible amplification of genetic material facilitating significant advancements in biomedical research
Catalog No. 50-008-97416 Supplier Apexbio Technology LLC Supplier No. K31022000
HyperScript IV Reverse Transcriptase is a pivotal enzyme in molecular biology research primarily used in the synthesis of complementary DNA (cDNA) from RNA templates Originating from retroviral sources this enzyme is crucial for studies involving gene expression analysis cloning and RNA sequencing It operates through a mechanism that reverses the transcription process effectively converting single-stranded RNA into stable cDNA Its activity is measured in the nanomolar to low micromolar range demonstrating high efficiency and reliability under in vitro conditions This enzyme is extensively applied in various experimental settings including cell models animal studies and high-throughput drug screening assays Experimental concentrations typically depend on the specific research design Using this advanced reverse transcriptase researchers can achieve precise and reproducible amplification of genetic material facilitating significant advancements in biomedical research