Learn More
Thermo Scientific™ HyperSep™ Universal Vacuum Manifold, Gauge and Waste Base
Catalog No. 03251251 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Universal Vacuum Manifold
Catalog No. For Use With (Equipment)
03-251-251 Universal Vacuum Manifold
1 options
Catalog No. 03-251-251 Supplier Thermo Scientific™ Supplier No. 60104231
Add to Cart
Add to Cart
Use this simple vacuum manifold for solid phase extraction (SPE) in 96 well plates.
Order the Thermo Scientific™ HyperSep™ Universal Vacuum Manifold, Gauge and Waste Base as a replacement spare part for your HyperSep Universal Vacuum Manifold. This manifold processes samples in solid phase extraction SPE cartridges or 96 well plate.
TRUSTED_SUSTAINABILITY
Specifications
Description Universal Vacuum Manifold, Gauge and Waste Base
For Use With (Application) SPE
For Use With (Equipment) Universal Vacuum Manifold
Product Line HyperSep
Type Gauge and Waste Base