Learn More
Thermo Scientific™ HyperSep™ Aminopropyl SPE 96-Well Plates and Individual Wells
Catalog No. 03251349 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Aminopropyl
Catalog No. Sorbent
03-251-349 HyperSep Aminopropyl
1 options
Catalog No. 03-251-349 Supplier Thermo Scientific™ Supplier No. 60300503
Obtain excellent retention of drugs, metabolites and structural isomers with these aminopropyl SPE well plates that allow both polar and anion exchange interactions.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Thermo Scientific™ HyperSep Aminopropyl SPE 96-Well Plates and Individual Wells feature a polar sorbent that allows both polar and anion exchange interactions. The chemistry offers excellent retention of drugs, metabolites and structural isomers. Typical applications include:
Separation of structural isomers
Drugs and metabolites in biological fluids
Saccharides, phenols and petroleum products
The sorbent used in HyperSep Aminopropyl SPE 96-Well Plates and Individual Wells acts as a weak anion exchanger, and also provides enhanced retention of strong anions like sulfonic acids.
HyperSep SPE Cartridges and Plates
Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE Wells; Bed weight 50 mg
For Use With (Application) Separation of Structural Isomers, Drugs and Metabolites in Biological Fluids
Particle Size 40 to 60 μm
Sorbent HyperSep Aminopropyl
Type Individual Well