Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ C8 Plates

Catalog No. 60307213 Shop All Thermo Scientific Products
Change view
Click to view available options
Bed Weight:
10 mg
25 mg
50 mg
100 mg
Volume (Metric) Well:
1 mL
2 mL
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Weight Volume (Metric) Well
60-307-213 50 mg 2 mL
60-307-211 10 mg 2 mL
60-307-212 25 mg 2 mL
03-251-331 10 mg 1 mL
03-251-332 25 mg 1 mL
03-251-333 50 mg 1 mL
03-251-334 100 mg 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 60-307-213 Supplier Thermo Scientific™ Supplier No. 60307213
Get an Extra Case of Product with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Need a less retentive alternative to C18 for polar and nonpolar compounds? These hydrophobic reversed phase C8 well plates offer excellent performance.

The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.

Thermo Scientific™ HyperSep™ C8 Plates offer a less retentive alternative to C18 for polar and nonpolar compounds, and are well-suited for methods requiring less retention than C18. The cartridges are ideally suited for the analysis of:

  • Drugs and their metabolites in biological samples
  • Peptides in biological samples

HyperSep C8 SPE Well Plates and Individual Wells provide excellent performance characteristics for moderately polar analytes. They also offer the potential for polar interactions greater than with C18.

HyperSep SPE Cartridges and Plates

HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.

The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.

TRUSTED_SUSTAINABILITY

Specifications

Description 96-Well Plate; Bed weight 50 mg/2 mL
Bed Weight 50 mg
For Use With (Application) Screening of Drugs, Metabolites, and Peptides in Biological Samples
Particle Size 40 to 60 μm
Product Line HyperSep
Sorbent HyperSep C8
Type Solid Phase Extraction Well Plate
Volume (Metric) Well 2 mL
Quantity 1 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.