Learn More
Thermo Scientific™ HyperSep™ Cyano Solid Phase Extraction (SPE) 96-Well Plates and Individual Wells
Catalog No. 03052233 Shop All Thermo Scientific Products
Change view
Click to view available options
Sorbent:
HyperSep Cyano
Catalog No. Sorbent
03-052-233 HyperSep Cyano
1 options
Catalog No. 03-052-233 Supplier Thermo Scientific™ Supplier No. 60300818
Get optimized isolation of polar compounds from nonpolar matrices, with these medium polarity cyano SPE well plates and individual wells.
The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
A medium polarity sorbent, Thermo Scientific™ HyperSep Cyano SPE 96-Well Plates and Individual Wells are less retentive than silica or diol, and are optimized for the isolation of polar compounds from nonpolar matrices. Typical applications include:
Retention of polar compounds from hexane and oils
Reversed phase extraction of moderately polar compounds
HyperSep SPE Cartridges and Plates
HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
TRUSTED_SUSTAINABILITY
Specifications
Description Individual SPE Wells; Bed weight 25 mg
For Use With (Application) Retention of Polar Compounds From Non-Polar Matrices
Particle Size 40 to 60 μm
Sorbent HyperSep Cyano
Type Individual Well